Learn More
IFN-γ Rat anti-Mouse, APC, Clone: XMG1.2, BD
Rat Monoclonal Antibody
103.79 € - 377.71 €
Specifications
| Antigen | IFN-γ |
|---|---|
| Clone | XMG1.2 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15811569
|
BD Biosciences
562018 |
25 μg |
104.00 € 103.79 € / Each Save 0.21 € 0.20% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15867678
|
BD Biosciences
554413 |
0.1 mg |
390.00 € 377.71 € / Each Save 12.29 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The XMG1.2 monoclonal antibody specifically binds to mouse interferon- γ(IFN- γ) protein. IFN- γis a pleiotropic cytokine, of approximately 15-17 kDa, involved in the regulation of inflammatory and immune responses. It plays an important role in activation, growth, and differentiation of T and B lymphocytes, macrophages, NK cells and other non-hematopoietic cell types. IFN- γproduction is associated with the Th1 cell differentiation. The purified form of this antibody has been reported to be a neutralizing antibody.
Specifications
| IFN-γ | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| IgG1 κ | |
| Affinity Purified |
| XMG1.2 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.