missing translation for 'onlineSavingsMsg'
Learn More

IFN-omega/IFNW1 Antibody, Novus Biologicals™

Product Code. 18642346 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18642346 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18642346 Supplier Novus Biologicals Supplier No. NBP299255100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFN-omega/IFNW1 Polyclonal antibody specifically detects IFN-omega/IFNW1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-omega/IFNW1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:1000-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias IFN-omega 1, interferon omega-1, Interferon alpha-II-1, interferon omega-1, interferon, omega 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human IFN-omega/IFNW1 (Uniprot#: P05000; Met1-Ser195)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3467
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.