missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-omega/IFNW1 Antibody (04), DyLight 488 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06154G
This item is not returnable.
View return policy
Description
IFN-omega/IFNW1 Monoclonal antibody specifically detects IFN-omega/IFNW1 in Human samples. It is validated for ELISA
Specifications
| IFN-omega/IFNW1 | |
| Monoclonal | |
| DyLight 488 | |
| IFN-omega 1, interferon omega-1, Interferon alpha-II-1, interferon omega-1, interferon, omega 1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IFN-omega/IFNW1 (P05000; Met1-Ser195). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 3467 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 04 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction