missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma Antibody (CC302) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22483-0.1MG
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human, Porcine, Bovine, Canine, Equine, Goat, Sheep samples. It is validated for Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| IFN-gamma | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000 | |
| P01579 | |
| IFNG | |
| Recombinant Bovine IFN gamma | |
| Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
| Human, Pig, Bovine, Canine, Equine, Goat, Sheep | |
| Purified |
| Flow Cytometry, ELISA | |
| CC302 | |
| Unconjugated | |
| PBS | |
| IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Mouse | |
| 0.1 mg | |
| 3458 | |
| Store at -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction