missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-beta Antibody (040), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89612IR
This item is not returnable.
View return policy
Description
IFN-beta Monoclonal antibody specifically detects IFN-beta in Human samples. It is validated for ELISA
Specifications
| IFN-beta | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 040 | |
| ELISA | |
| Fibroblast interferon, IFBIFNB, IFF, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN-beta (NP_002167.1; Met1-Asn187). | |
| 0.1 mL | |
| Apoptosis, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3456 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction