missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IFIT3 Polyclonal antibody specifically detects IFIT3 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | IFIT3 |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CIG-49, GARG-49, IFI60Retinoic acid-induced gene G protein, IFIT-3, IFIT-4, IFIT4IFI-60K, interferon-induced protein with tetratricopeptide repeats 3, Interferon-induced protein with tetratricopeptide repeats 4Interferon-induced 60 kDa protein, IRG2, ISG60, ISG-60, RIG-GCIG49 |
| Host Species | Rabbit |
| Immunogen | IFIT3 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human IFIT3. The immunogen is located within amino acids 380 - 430 of IFIT3 . |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?