missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFI27 Monoclonal Antibody (CL14627)

Product Code. 30829965
Change view
Click to view available options
Quantity:
100 μL
Product Code. Quantity
30829965 100 μL
1 options
This item is not returnable. View return policy

Product Code. 30829965

Brand: Invitrogen™ MA562982

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFI27 Monoclonal Antibody for IHC (P)

Promotes cell death. Mediates IFN-induced apoptosis characterized by a rapid and robust release of cytochrome C from the mitochondria and activation of BAX and caspases 2, 3, 6, 8 and 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFI27
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL14627
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFI27
Gene Accession No. P40305
Gene Alias 2310061N23Rik; FAM14D; IFI27; interferon alpha inducible protein 27; interferon alpha-induced 11.5 kDa protein; Interferon alpha-inducible protein 27, mitochondrial; interferon, alpha-inducible protein 27; interferon-stimulated gene 12a protein; ISG12; ISG12(a); ISG12A; p27
Gene Symbols IFI27
Host Species Mouse
Immunogen Recombinant protein corresponding to Human IFI27.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3429
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.