Learn More
Fisherbrand™ IEF Conversion Kit
Product Code. 15116634 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Verti-Gel Maxi 2D
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15116634 Verti-Gel Maxi 2D Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15116634
Only null left
Add to basket
Add to basket
Accessory for Verti-Gel Maxi 2D Electrophoresis System.
  • For 18cm IPG strips and tube gels
  • For 2D runs with 180mm IPG strips and gels
Specifications
Description For 2D runs with 180mm IPG strips and gels
For Use With (Equipment) Verti-Gel Maxi 2D
Type IEF Conversion Kit
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.