missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IdU Monoclonal Antibody (OTI2B10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524879
This item is not returnable.
View return policy
Description
IdU Monoclonal Antibody for ICC/IF, IHC (P)
Idu (Idoxuridine) is a nucleoside analogue, a modified form of deoxyuridine, similar enough to be incorporated into viral DNA replication, but the iodine atom added to the uracil component blocks base pairing. It is used only topically due to cardiotoxicity. It was synthesized by William Prusoff in the late 1950s.
Specifications
| IdU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| OTI2B10 | |
| Unconjugated | |
| IdU | |
| Iododeoxyuridine coupled to keyhole limpet hemocyanin. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction