Learn More
IDS Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29375-1-AP-20UL
Description
The IDS antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IDS. This antibody recognizes human, mouse, rat antigen. The IDS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IDS | |
| 700 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| IDS, iduronate 2 sulfatase, Idursulfase, MPS2, SIDS | |
| 20 μL | |
| 3423 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.