missing translation for 'onlineSavingsMsg'
Learn More

IDO-IN-3, MedChemExpress

Product Code. p-200100869
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497238 10 mg 10mg
16479767 5 mg 5mg
16463257 50 mg 50mg
16400296 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497238 Supplier Medchem Express Supplier No. HY16987
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress IDO-IN-3 is a potent indoleamine 2,3-dioxygenase (IDO) inhibitor with an IC50 of 290 nM.

IDO-IN-3 is a potent indoleamine 2,3-dioxygenase (IDO) inhibitor with an IC50 of 290 nM.

Specifications

Chemical Name or Material IDO-IN-3
CAS 2070018-30-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H12BrFN6O2
Quantity 10 mg
Solubility Information DMSO : 27.5 mg/mL (76.57 mM; Need ultrasonic and warming)
SMILES O/N=C(NC1=CC(Br)=C(F)C=C1)\C2=NON=C2NCCN
Molecular Weight (g/mol) 359.15
Formula Weight 359.15
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.