Description
- Durable and reusable foam stoppers are an economical way to keep the contents of your containers secure.
- Open-cell foam has a uniform density.
- Leaves no fibers like cotton
- Breathable, nonabsorbent, chemcial resistant, heat tolerant, and retains shape after repeated use.
- Can also be used as a cleaner for pipettes, tubes, vials and precision tubing.
- Autoclavable.
Specifications
Specifications
| Diameter (Metric) Inner | 27 to 34 mm |
| For Use With (Application) | Cleaner for pipettes, tubes, vials and precision tubing |
| Height (Metric) | 44.5 mm |
| Quantity | 200 Pk. |
| Autoclavable | Autoclavable |
| Color | White |
| Material | Plastic |
| Product Type | Plug Stopper |