missing translation for 'onlineSavingsMsg'
Learn More

Idalopirdine Hydrochloride, MedChemExpress

Product Code. 16442177
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442177

Brand: Medchem Express HY14338A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Idalopirdine Hydrochloride (Lu AE58054 Hydrochloride) is a potent and selective 5-HT6 receptor antagonist with a Ki of 0.83 nM.

Idalopirdine Hydrochloride (Lu AE58054 Hydrochloride) is a potent and selective 5-HT6 receptor antagonist with a Ki of 0.83 nM.

Specifications

Chemical Name or Material Idalopirdine Hydrochloride
CAS 467458-02-2
Color Gray
Purity Grade Notes Research
Molecular Formula C20H20ClF5N2O
Quantity 50 mg
Synonym Lu AE58054 Hydrochloride
Solubility Information DMSO : ≥ 25 mg/mL (57.49 mM) ∣H2O : 2 mg/mL (4.60 mM; ultrasonic and warming and heat to 60°C)
SMILES FC1=CC(NC=C2CCNCC3=CC(OCC(F)(F)C(F)F)=CC=C3)=C2C=C1.Cl
Molecular Weight (g/mol) 434.83
Formula Weight 434.83
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.