Learn More
Idalopirdine Hydrochloride, MedChemExpress
Product Code. 16498637
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16498637
Brand: Medchem Express HY14338A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Idalopirdine Hydrochloride (Lu AE58054 Hydrochloride) is a potent and selective 5-HT6 receptor antagonist with a Ki of 0.83 nM.
Idalopirdine Hydrochloride (Lu AE58054 Hydrochloride) is a potent and selective 5-HT6 receptor antagonist with a Ki of 0.83 nM.
Specifications
Chemical Name or Material Idalopirdine Hydrochloride
CAS 467458-02-2
Color Gray
Purity Grade Notes Research
Molecular Formula C20H20ClF5N2O
Quantity 5 mg
Synonym Lu AE58054 Hydrochloride
Solubility Information DMSO : ≥ 25 mg/mL (57.49 mM) ∣H2O : 2 mg/mL (4.60 mM; ultrasonic and warming and heat to 60°C)
SMILES FC1=CC(NC=C2CCNCC3=CC(OCC(F)(F)C(F)F)=CC=C3)=C2C=C1.Cl
Molecular Weight (g/mol) 434.83
Formula Weight 434.83
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation