Learn More
Invitrogen™ ID4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540522
Description
Peptide sequence: CYSRLRRLVP TIPPNKKVSK VEILQHVIDY ILDLQLALET HPALLRQPPP Sequence homology: Cow: 85%; Dog: 100%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 79%; Rat: 100%; Zebrafish: 86%.
ID4 is transcription factors containing a basic helix-loop-helix (bHLH) motif regulate expression of tissue-specific genes in a number of mammalian and insect systems. DNA-binding activity of the bHLH proteins is dependent on formation of homo- and/or heterodimers. Dominant-negative HLH proteins encoded by Id-related genes, such as ID4, also contain the HLH-dimerization domain but lack the DNA-binding basic domain. Consequently, Id proteins inhibit binding to DNA and transcriptional transactivation by heterodimerization with bHLH proteins.
Specifications
| ID4 | |
| Polyclonal | |
| Unconjugated | |
| Id4 | |
| bHLHb27; cb547; Class B basic helix-loop-helix protein 27; DNA-binding protein inhibitor ID-4; fb77d04; hypothetical protein LOC664761; ID4; Id-4; Idb4; Inhibitor of differentiation 4; inhibitor of DNA binding 4; inhibitor of DNA binding 4, dominant negative helix-loop-helix protein; inhibitor of DNA binding 4, HLH protein; sb:cb547; wu:fb77d04; zgc:123214 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3400 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P47928 | |
| Id4 | |
| Synthetic peptide directed towards the middle region of human ID4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.