Learn More
Sampling Systems™ ID Tape
Product Code. 15879501 Shop All Sampling Systems Products
Click to view available options
Display Text:
Quarantine
Rejected
Released
Sampled
Unit Size:
10 rolls
Pack of 10
Product Code. 15879501
Brand: Sampling Systems™ 8221A03
Only null left
Add to basket
Add to basket
Quickly identify the status of a container to prevent it being used in error. Sampling Systems™ ID Tape allows containers, pallets, and bags to be easily labelled with highly visible tape.
  • Use with standard tape dispensers
  • High strength adhesive
  • 10 rolls per box
Specifications
Description Medium strength adhesion tape
Color Red
Product Type Labeled ID Tape
Display Text Released
No. per Pack 10/Pk.
Dimensions (LxW) 66 x 50 mm
Length (Metric) 66 mm
Width (Metric) 50 mm