Learn More
iCRT3, MedChemExpress
Product Code. 16496617
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496617 5 mg 5mg
16492896 25 mg 25mg
16463898 10 mg 10mg
16444936 100 mg 100mg
16400267 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16496617 Supplier Medchem Express Supplier No. HY103705
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress iCRT3 is an inhibitor of both Wnt and β-catenin-responsive transcription.
iCRT3 is an inhibitor of both Wnt and β-catenin-responsive transcription.
Specifications
Chemical Name or Material iCRT3
Physical Form Solid
CAS 901751-47-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H26N2O2S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 150 mg/mL (380.20 mM; Need ultrasonic and warming)
SMILES O=C(NCCC1=CC=CC=C1)CSCC2=C(C)OC(C3=CC=C(CC)C=C3)=N2
Molecular Weight (g/mol) 394.53
Formula Weight 394.53
Percent Purity 98.18%
Grade Research