Learn More
ICI 63197, TRC
Product Code. 30400476
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400476 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30400476 Supplier Toronto Research Chemicals Supplier No. TRCI1638885MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material ICI 63197
CAS 27277-00-5
Molecular Formula C9H13N5O
Quantity 5 mg
Synonym 2-Amino-6-methyl-4-propyl-[1,2,4]triazolo[1,5-a]pyrimidin-5(4H)-one, 2-Amino-4,5-dihydro-6-methyl-4-propyl-s-triazolo[1,5-a]pyrimidin-5-one, 2-Amino-6-methyl-4-propyl-4H-[1,2,4]triazolo[1,5-a]pyrimidin-5-one, 2-Amino-6-methyl-4-propyl-s-triazolo[1,5-a]pyrimid-5(4H)-one, 2-Amino-6-methyl-5-oxo-4-propyl-4,5-dihydro-s-triazolo[1,5-a]pyrimidine
InChI Formula InChI=1S/C9H13N5O/c1-3-4-13-7(15)6(2)5-14-9(13)11-8(10)12-14/h5H,3-4H2,1-2H3,(H2,10,12)
SMILES CCCN1C2=NC(N)=NN2C=C(C1=O)C
IUPAC Name 2-amino-6-methyl-4-propyl-[1,2,4]triazolo[1,5-a]pyrimidin-5-one
Molecular Weight (g/mol) 207.23
Formula Weight 207.112