Learn More
ICI 63197, TRC
Product Code. 30398275
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398275 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30398275 Supplier Toronto Research Chemicals Supplier No. TRCI16388825MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material ICI 63197
CAS 27277-00-5
Molecular Formula C9H13N5O
Quantity 25 mg
Synonym 2-Amino-6-methyl-4-propyl-[1,2,4]triazolo[1,5-a]pyrimidin-5(4H)-one, 2-Amino-4,5-dihydro-6-methyl-4-propyl-s-triazolo[1,5-a]pyrimidin-5-one, 2-Amino-6-methyl-4-propyl-4H-[1,2,4]triazolo[1,5-a]pyrimidin-5-one, 2-Amino-6-methyl-4-propyl-s-triazolo[1,5-a]pyrimid-5(4H)-one, 2-Amino-6-methyl-5-oxo-4-propyl-4,5-dihydro-s-triazolo[1,5-a]pyrimidine
InChI Formula InChI=1S/C9H13N5O/c1-3-4-13-7(15)6(2)5-14-9(13)11-8(10)12-14/h5H,3-4H2,1-2H3,(H2,10,12)
SMILES CCCN1C2=NC(N)=NN2C=C(C1=O)C
IUPAC Name 2-amino-6-methyl-4-propyl-[1,2,4]triazolo[1,5-a]pyrimidin-5-one
Molecular Weight (g/mol) 207.23
Formula Weight 207.112