missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Icarapin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144566
This item is not returnable.
View return policy
Description
Icarapin Polyclonal Antibody for Western Blot, ELISA
Icarapin is a peptide that is derived from the venom of the South American pit viper, Bothrops jararaca. It has been studied for its potential therapeutic properties, particularly its antimicrobial and antifungal activities. Icarapin has shown inhibitory effects against a range of bacteria and fungi, including drug-resistant strains. Research is ongoing to further explore the potential applications and mechanisms of action of icarapin, as well as its potential as a source for the development of new antimicrobial agents.
Specifications
| Icarapin | |
| Polyclonal | |
| Unconjugated | |
| Icarapin | |
| Recombinant Apis mellifera carnica Icarapin protein (20-223aa). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| Insect | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction