Learn More
Invitrogen™ ICAM-3 Monoclonal Antibody (ICAM3.1)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA190977
Product Code. 11551062
  • 582.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD50 (intracellular adhesion molecule 3, ICAM-3) is a transmembrane glycoprotein expressed by leukocytes, that serves as a counter-receptor for the lymphocyte function-associated antigen (LFA)-1 integrin. Besides functioning as an adhesive molecule that mediates e.g. the contact between T cells and antigen presenting cells, ICAM-3 regulates affinity of LFA-1 for ICAM-1 and induces T cell activation and proliferation. ICAM-3 plays an essential role in the initiation of the immune response both on T cells and antigen presenting cells and interacts also with CD209 (dendritic cell-specific ICAM-3-grabbing nonintegrin, DC-SIGN), a C-type lectin of dendritic cells and macrophages; this process is involved in dialogue between dendritic cells and granulocytes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ICAM-3
Monoclonal
1 mg/mL
PBS with no preservative
P32942
ICAM3
ICAM 3Fc chimeric fusion protein.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
ICAM3.1
Unconjugated
ICAM3
CD50; CDw50; ICAM3; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3
Mouse
Protein A
RUO
3385
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.