missing translation for 'onlineSavingsMsg'
Learn More

Ibutilide Fumarate, TRC

Product Code. 30007760
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007760 100 mg 100mg
30135617 10 mg 10mg
30134899 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007760 Supplier Toronto Research Chemicals Supplier No. TRCI155400100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122647-32-9
Formula Weight 884.5003
InChI Formula InChI=1S/2C20H36N2O3S.C4H4O4/c2*1-4-6-7-8-9-16-22(5-2)17-10-11-20(23)18-12-14-19(15-13-18)21-26(3,24)25;5-3(6)1-2-4(7)8/h2*12-15,20-21,23H,4-11,16-17H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+
IUPAC Name (E)-but-2-enedioic acid;N-[4-[4-[ethyl(heptyl)amino]-1-hydroxybutyl]phenyl]methanesulfonamide
Molecular Formula 2 C20 H36 N2 O3 S . C4 H4 O4
Molecular Weight (g/mol) 885.22
SMILES CCCCCCCN(CC)CCCC(O)c1ccc(NS(=O)(=O)C)cc1.CCCCCCCN(CC)CCCC(O)c2ccc(NS(=O)(=O)C)cc2.OC(=O)\C=C\C(=O)O
Synonym Methanesulfonamide, N-[4-[4-(ethylheptylamino)-1-hydroxybutyl]phenyl]-, (2E)-2-butenedioate (2:1), Methanesulfonamide, N-[4-[4-(ethylheptylamino)-1-hydroxybutyl]phenyl]-, (2E)-2-butenedioate (2:1) (salt) (9CI), Methanesulfonamide, N-[4-[4-(ethylheptylamino)-1-hydroxybutyl]phenyl]-, (±)-, (E)-2-butenedioate (2:1) (salt), Corvert, Ibutilide fumarate, Methanesulfonamide, N-[4-[4-(ethylheptylamino)-1-hydroxybutyl]phenyl]-, (E)-2-butenedioate (2:1) (salt), U 70226E
Chemical Name or Material Ibutilide Fumarate

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.