Learn More
Ibuprofen piconol, MedChemExpress
Product Code. 16449916
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Product Code. 16449916
Brand: Medchem Express HY101482
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Ibuprofen piconol is a non-steroidal, anti-inflammatory (NSAID) agent for the topical relief of primary thermal burns and sunburns.
Ibuprofen piconol is a non-steroidal, anti-inflammatory (NSAID) agent for the topical relief of primary thermal burns and sunburns.
Specifications
Chemical Name or Material Ibuprofen piconol
Physical Form Liquid
CAS 64622-45-3
Color Colorless
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H23NO2
For Use With (Application) COVID-19-immunoregulation
Synonym U75630
Solubility Information DMSO : ≥ 50 mg/mL (168.13 mM)
SMILES O=C(OCC1=NC=CC=C1)C(C)C2=CC=C(CC(C)C)C=C2
Molecular Weight (g/mol) 297.39
Formula Weight 297.39
Percent Purity 98.0%
Grade Research