missing translation for 'onlineSavingsMsg'
Learn More

IBR2, MedChemExpress

Product Code. p-200096359
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464936 100 mg 100mg
16436627 5 mg 5mg
16430267 50 mg 50mg
16403908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464936 Supplier Medchem Express Supplier No. HY103710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IBR2 is a potent and specific RAD51 inhibitor and inhibits RAD51-mediated DNA double-strand break repair. IBR2 disrupts RAD51 multimerization, accelerates proteasome-mediated RAD51 protein degradation, inhibits cancer cell growth and induces apoptosis.

IBR2 is a potent and specific RAD51 inhibitor and inhibits RAD51-mediated DNA double-strand break repair. IBR2 disrupts RAD51 multimerization, accelerates proteasome-mediated RAD51 protein degradation, inhibits cancer cell growth and induces apoptosis.

Specifications

Chemical Name or Material IBR2
CAS 313526-24-8
Color Pink
Purity Grade Notes Research
Molecular Formula C24H20N2O2S
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (249.69 mM)
SMILES O=S(N1C(C2=CNC3=C2C=CC=C3)C4=C(C=CC=C4)C=C1)(CC5=CC=CC=C5)=O
Molecular Weight (g/mol) 400.49
Formula Weight 400.49
Percent Purity 98.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.