missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IARS Polyclonal Antibody
GREENER_CHOICE

Product Code. 17710644
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17710644 200 μL 200µL
17753234 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17710644 Supplier Invitrogen™ Supplier No. PA5119234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IARS Polyclonal Antibody for IHC (P), Western Blot

IARS is a member of aminoacyl-tRNA synthetases which catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAS, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Isoleucine-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family and has been identified as a target of autoantibodies in the autoimmune disease polymyositis/dermatomyositis. Two alternatively spliced variants have been isolated that represent alternate 5' UTRs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IARS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene IARS1
Gene Accession No. P41252
Gene Alias 2510016L12Rik; AI327140; AU044614; E430001P04Rik; fi46h05; FLJ20736; IARS; IARS1; Iarsl; ILERS; ILRS; IRS; isoleucine tRNA ligase 1, cytoplasmic; isoleucine tRNA synthetase; isoleucine--tRNA ligase, cytoplasmic; isoleucine-tRNA synthetase; isoleucyl tRNA synthetase; isoleucyl-tRNA synthetase; isoleucyl-tRNA synthetase 1; isoleucyl-tRNA synthetase, cytoplasmic; PRO0785; wu:fi46h05; zgc:63790
Gene Symbols IARS1
Host Species Rabbit
Immunogen Recombinant protein Isoleucyl tRNA Synthetase. The antigen corresponds to amino acid range 1-500 of the target protein.
Purification Method Protein A, Antigen affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3376
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.