Learn More
Iadademstat dihydrochloride, MedChemExpress
Product Code. 16481537
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16481537
Brand: Medchem Express HY12782T
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Iadademstat (ORY-1001) dihydrochloride is a selective irreversible lysine (K)-specific demethylase 1A (KDM1A/LSD1) inhibitor.
Iadademstat (ORY-1001) dihydrochloride is a selective irreversible lysine (K)-specific demethylase 1A (KDM1A/LSD1) inhibitor.
Specifications
Chemical Name or Material Iadademstat dihydrochloride
Physical Form Solid
CAS 1431303-72-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H24Cl2N2
For Use With (Application) Cancer-programmed cell death
Synonym ORY-1001 dihydrochlorideRG6016 dihydrochlorideRO 7051790 dihydrochloride
Solubility Information H2O : 57.75 mg/mL (190.42 mM; Need ultrasonic and warming) ∣DMSO : 0.69 mg/mL (2.28 mM; Need ultrasonic and warming)
SMILES N[C@@H]1CC[C@@H](N[C@H]2[C@H](C3=CC=CC=C3)C2)CC1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 303.27
Formula Weight 303.27
Percent Purity 98.3%
Grade Research