Learn More
The Central Surgical Company™ Hypodermic Syringe Needles
Click to view available options
Needle Gauge:
18 G
20 G
22 G
Length (Metric) Needle:
32 mm
41 mm
50 mm
Unit Size:
Pack of 10
Specifications
Specifications
| Needle Gauge | 18 G |
| Product Type | Hypodermic Syringe Needle |
| Length (Metric) Needle | 50 mm |