missing translation for 'onlineSavingsMsg'
Learn More

Hypidone hydrochloride, MedChemExpress

Product Code. p-200097578
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493158

Brand: Medchem Express HY100769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hypidone hydrochloride (YL0919) is an orally active antidepressant agent with dual activity as a highly seletive 5-HT uptake blocker and an effective 5-HT1A receptor agonist (Ki=0.19 nM).

Hypidone hydrochloride inhibits the uptake of [3H]-5-HT into rat cerebral cortical synaptosomes and HEK293 cells with IC50s of 1.78 nM and 1.93 nM, respectively. Hypidone hydrochloride shows remarkable antidepressant effects in animal models and has the poential for the investigation of depressive disorder.

Specifications

Chemical Name or Material Hypidone hydrochloride
CAS 1339058-04-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H23ClN2O2
Quantity 10 mg
Synonym YL0919
Solubility Information DMSO : ≥ 30 mg/mL (89.60 mM)
SMILES O=C1C=CC=CN1CC2(O)CCN(CC3=CC=CC=C3)CC2.[H]Cl
Molecular Weight (g/mol) 334.84
Formula Weight 334.84
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.