Learn More
Hypidone hydrochloride, MedChemExpress
Product Code. 16493158
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Product Code. 16493158
Brand: Medchem Express HY100769
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Hypidone hydrochloride (YL0919) is an orally active antidepressant agent with dual activity as a highly seletive 5-HT uptake blocker and an effective 5-HT1A receptor agonist (Ki=0.19 nM).
Hypidone hydrochloride inhibits the uptake of [3H]-5-HT into rat cerebral cortical synaptosomes and HEK293 cells with IC50s of 1.78 nM and 1.93 nM, respectively. Hypidone hydrochloride shows remarkable antidepressant effects in animal models and has the poential for the investigation of depressive disorder.
Specifications
Chemical Name or Material Hypidone hydrochloride
CAS 1339058-04-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H23ClN2O2
Quantity 10 mg
Synonym YL0919
Solubility Information DMSO : ≥ 30 mg/mL (89.60 mM)
SMILES O=C1C=CC=CN1CC2(O)CCN(CC3=CC=CC=C3)CC2.[H]Cl
Molecular Weight (g/mol) 334.84
Formula Weight 334.84
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation