Learn More
Hydroxy Donepezil(Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | Hydroxy Donepezil (Mixture of Diastereomers) |
| CAS | 197010-20-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 H29 N O4 |
| Quantity | 25 mg |
| Synonym | 1H-Inden-1-one, 2,3-dihydro-2-[hydroxy[1-(phenylmethyl)-4-piperidinyl]methyl]-5,6-dimethoxy-, 2-[(1-Benzylpiperidin-4-yl)hydroxymethyl]-5,6-dimethoxyindan-1-one, Hydroxy Donepezil, Hydroxydonepezil |
| InChI Formula | InChI=1S/C24H29NO4/c1-28-21-13-18-12-20(24(27)19(18)14-22(21)29-2)23(26)17-8-10-25(11-9-17)15-16-6-4-3-5-7-16/h3-7,13-14,17,20,23,26H,8-12,15H2,1-2H3 |
| SMILES | COc1cc2CC(C(O)C3CCN(Cc4ccccc4)CC3)C(=O)c2cc1OC |
| IUPAC Name | 2-[(1-benzylpiperidin-4-yl)-hydroxymethyl]-5,6-dimethoxy-2,3-dihydroinden-1-one |
| Molecular Weight (g/mol) | 395.4914 |
| Show More |