Learn More
Hydroxy Donepezil(Mixture of Diastereomers), TRC
Product Code. 30391415
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
missing translation for 'unitSize'
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391415 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Product Code. 30391415 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCH941740100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Hydroxy Donepezil (Mixture of Diastereomers)
CAS 197010-20-1
Purity Grade Notes HPLC
Molecular Formula C24 H29 N O4
Quantity 100 mg
Synonym 1H-Inden-1-one, 2,3-dihydro-2-[hydroxy[1-(phenylmethyl)-4-piperidinyl]methyl]-5,6-dimethoxy-, 2-[(1-Benzylpiperidin-4-yl)hydroxymethyl]-5,6-dimethoxyindan-1-one, Hydroxy Donepezil, Hydroxydonepezil
InChI Formula InChI=1S/C24H29NO4/c1-28-21-13-18-12-20(24(27)19(18)14-22(21)29-2)23(26)17-8-10-25(11-9-17)15-16-6-4-3-5-7-16/h3-7,13-14,17,20,23,26H,8-12,15H2,1-2H3
SMILES COc1cc2CC(C(O)C3CCN(Cc4ccccc4)CC3)C(=O)c2cc1OC
IUPAC Name 2-[(1-benzylpiperidin-4-yl)-hydroxymethyl]-5,6-dimethoxy-2,3-dihydroinden-1-one
Molecular Weight (g/mol) 395.4914
Formula Weight 395.2097
Percent Purity >95