Learn More
Merck Hydrosol Stainless Steel Filter Holder and Accessories
Accessories for filter holders for sample preparation.
515.00 € - 1785.00 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
For Use With (Equipment) | 47 mm hydrosol vacuum filter |
Sterility | Non-sterile |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10111541
|
Merck
XX20047RK |
Replacement Parts Kit |
515.00 €
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
10654552
|
Merck
XX2004704 |
Funnel and Locking Ring Assembly |
1785.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
10450103
|
Merck
XX2004702 |
Stainless steel base and filter screen, silicone stopper, PTFE gasket |
1200.00 €
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
Accessories for filter holders for sample preparation.
The Hydrosol filter holder replacement PartsXX2004704 Funnel and Locking Ring Assembly, stainless steelXX2004702 Base with Screen, gasket and supportXX20047RK Replacement Parts Kit for Hydrosol Filter HolderSpecifications
Not Autoclavable | |
47 mm hydrosol vacuum filter | |
Non-sterile |