Learn More
Thermo Scientific™ Hydrogold Column Guard
Product Code. 15520312 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
2 m
5 m
Unit Size:
Each
Product Code. 15520312
Brand: Thermo Scientific™ 26H500253
Only null left
Add to basket
Add to basket
Provides protection to the analytical column.
Protects against column contamination caused by non-volatile materials, extending the column lifetime.
TRUSTED_SUSTAINABILITY
Specifications
Type GC Guard Column
Diameter (Metric) Inner 0.53 mm
Length (Metric) 2 m
Quantity 1