Learn More
Hydrocortisone 17-Valerate, TRC
Product Code. 30407197
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407197 1 g 1g
30396327 10 g -
30405191 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30407197 Supplier Toronto Research Chemicals Supplier No. TRCH7147401G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57524-89-7
Molecular Formula C26 H38 O6
Molecular Weight (g/mol) 446.58
Synonym (11β)-11,21-Dihydroxy-17-[(1-oxopentyl)oxy]-pregn-4-ene-3,20-dione, Cortisol, 17-valerate (7CI), 11β,21-Dihydroxy-17α-valeryloxy-4-pregnene-3,20-dione, HydroVal, Hydrocortisone 17-valerate, Hydrocortisone pentanoate, Hydrocortisone valerate, Westcort
IUPAC Name [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] pentanoate
SMILES CCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO
Specifications
Quantity 1 g
InChI Formula InChI=1S/C26H38O6/c1-4-5-6-22(31)32-26(21(30)15-27)12-10-19-18-8-7-16-13-17(28)9-11-24(16,2)23(18)20(29)14-25(19,26)3/h13,18-20,23,27,29H,4-12,14-15H2,1-3H3/t18-,19-,20-,23+,24-,25-,26-/m0/s1
Formula Weight 446.2668
Chemical Name or Material Hydrocortisone Valerate