missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum, Australian Origin
Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA)
Brand: Cytiva SH30545.04HI
Additional Details : Weight : 1.15000kg
Description
- Sourced from Australian abattoirs approved by USDA for export and inspected by the Australian Department of Agriculture (DA)
- Australian methods of animal husbandry exhibit excellent animal nutrition and healthcare
- Animal disease control and management in Australia is easier compared to most areas of the world due to isolation
- Filtered through triple 100 nm pore size-rated filters
- Standard endotoxin and hemoglobin specifications
- Subjected to moderate biochemical analyses
Specifications
Australia | |
Fetal Bovine Serum | |
Sequential Triple-filtered, 100 nm | |
1000 mL | |
HyClone™ |
Serum | |
1000 mL | |
Heat Inactivated | |
Yes | |
European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests |