Learn More
Hyaluronidase, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Hyaluronidase (Hyaluronate 4-glycanohydrolase
Specifications
Specifications
| Chemical Name or Material | Hyaluronidase |
| Physical Form | Powder |
| CAS | 37326-33-3 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Synonym | Hyaluronate 4-glycanohydrolase Hyaluronoglucosaminidase |
| Solubility Information | H2O : 100 mg/mL (Need ultrasonic) |
| SMILES | [Hyaluronoglucosaminidase] |
| Grade | Research |