missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HVEM/TNFRSF14 Antibody (HMHV-1B18) - Chimeric - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
HVEM/TNFRSF14 Monoclonal antibody specifically detects HVEM/TNFRSF14 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Neutralization, Functional Assay
Specifications
Specifications
| Antigen | HVEM/TNFRSF14 |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Neutralization, Functional Assay |
| Classification | Monoclonal |
| Clone | HMHV-1B18 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 |
| Host Species | Armenian Hamster |
| Immunogen | This antibody was raised by immunising Armenian hamsters with mouse HVEM:Fc fusion protein. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?