missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HUWE1 Antibody (BLR129H), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14712
This item is not returnable.
View return policy
Description
HUWE1 Monoclonal antibody specifically detects HUWE1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| HUWE1 | |
| Monoclonal | |
| Unconjugated | |
| Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA and 0.09% Sodium Azide | |
| Rabbit | |
| 100 μg | |
| Stem Cell Markers | |
| 10075 | |
| Store at 2 - 8°C/ 1 year from date of receipt |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| BLR129H | |
| Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 20 ul/1 mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| arf-bp1, EC 6.3.2, EC 6.3.2.6, HECT, UBA and WWE domain containing 1, HECTH9, HSPC272, KIAA0312MULE, Mcl-1 ubiquitin ligase E3, Mule, UREB1UBA and WWE domain-containing protein 1, URE-binding protein 1 | |
| between 2250 and 2300 | |
| RUO | |
| Primary | |
| Human, Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction