missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HuR Polyclonal Antibody

Description
This antibody is predicted to react with chicken and cow based on sequence homology. Store antibody at 4ºC for 1-2 weeks. For long-term storage, store at -20ºC.
The protein encoded by this gene is a member of the ELAVL protein family. This encoded protein contains 3 RNA-binding domains and binds cis-acting AU-rich elements. It destabilizes mRNAs and thereby regulates gene expression.
Specifications
Specifications
| Antigen | HuR |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | ELAVL1 |
| Gene Accession No. | P70372, Q15717 |
| Gene Alias | 2410055N02Rik; ELAV (embryonic lethal, abnormal vision)-like 1 (Hu antigen R); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R); ELAV like 1; ELAV like RNA binding protein 1; ELAV like RNA binding protein 1a; ELAV1; Elavl1; elavl1 protein; elavl1a; elav-like generic protein; ELAV-like protein 1; ELAV-like protein 1; ELAV-like protein 1a; elav-related A; Elra; embryonic lethal, abnormal vision, drosophila, homolog-like 1; embryonic lethal, abnormal vision-related A; etID309757.2; fa25f08; fb59a02; fc60g02; Hu antigen R; Hua; HU-antigen A; hu-antigen R; Human antigen R; HuR; melG; ribonucleoprotein; W91709; wu:fa25f08; wu:fb59a02; wu:fc60g02; zHuA |
| Gene Symbols | ELAVL1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?