Learn More
Abnova™ Human ZP2 Full-length ORF (AAH96305.1, 1 a.a. - 736 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAH96305.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7783 |
Molecular Weight (g/mol) | 107.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16182112
|
Abnova™
H00007783-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16172112
|
Abnova™
H00007783-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The zona pellucida is an extracellular matrix that surrounds the oocyte and early embryo. It is composed primarily of three or four glycoproteins with various functions during fertilization and preimplantation development. The protein encoded by this gene is a structural component of the zona pellucida and functions in secondary binding and penetration of acrosome-reacted spermatozoa. The nascent protein contains a N-terminal signal peptide sequence, a conserved ZP domain, a consensus furin cleavage site, and a C-terminal transmembrane domain. It is hypothesized that furin cleavage results in release of the mature protein from the plasma membrane for subsequent incorporation into the zona pellucida matrix. However, the requirement for furin cleavage in this process remains controversial based on mouse studies. [provided by RefSeq]
Sequence: MACRQRGGSWSPSGWFNAGWSTYRSISLFFALVTSGNSIDVSQLVNPAFPGTVTCDEREITVEFPSSPGTKKWHASVVDPLGLDMPNCTYILDPEKLTLRATYDNCTRRVHGGHQMTIRVMNNSAALRHGAVMYQFFCPAMQVEETQGLSASTICQKDFMSFSLPRVFSGLADDSKGTKVQMGWSIEVGDGARAKTLTLPEAMKEGFSLLIDNHRMTFHVPFNATGVTHYVQGNSHLYMVSLKLTFISPGQKVIFSSQAICAPDPVTCNATHMTLTIPEFPGKLKSVSFENQNIDVSQLHDNGIDLEATNGMKLHFSKTLLKTKLSEKCLLHQFYLASLKLTFLLRPETVSMVIYPECLCESPVSIVTGELCTQDGFMDVEVYSYQTQPALDLGTLRVGNSSCQPVFEAQSQGLVRFHIPLNGCGTRYKFEDDKVVYENEIHALWTDFPPSKISRDSEFRNDMLLNINVESLTPPVASVKLGPFTLILQSYPDNSYQQPYGENEYPLVRFLRQPIYMEVRVLNRDDPNIKLVLDDCWATSTMDPDSFPQWNVVVDGCAYDLDNYQTTFHPVGSSVTHPDHYQRFDMKAFAFVSEAHVLSSLVYFHCSALICNRLSPDSPLCSVTCPVSSRHRRATGATEAEKMTVSLPGPILLLSDDSSFRGVGSSDLKASGSSGEKSRSETGEEVGSRGAMDTKGHKTAGDVGSKAVAAVAAFAGVVATLGFIYYLYEKRTVSNHSpecifications
AAH96305.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
107.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MACRQRGGSWSPSGWFNAGWSTYRSISLFFALVTSGNSIDVSQLVNPAFPGTVTCDEREITVEFPSSPGTKKWHASVVDPLGLDMPNCTYILDPEKLTLRATYDNCTRRVHGGHQMTIRVMNNSAALRHGAVMYQFFCPAMQVEETQGLSASTICQKDFMSFSLPRVFSGLADDSKGTKVQMGWSIEVGDGARAKTLTLPEAMKEGFSLLIDNHRMTFHVPFNATGVTHYVQGNSHLYMVSLKLTFISPGQKVIFSSQAICAPDPVTCNATHMTLTIPEFPGKLKSVSFENQNIDVSQLHDNGIDLEATNGMKLHFSKTLLKTKLSEKCLLHQFYLASLKLTFLLRPETVSMVIYPECLCESPVSIVTGELCTQDGFMDVEVYSYQTQPALDLGTLRVGNSSCQPVFEAQSQGLVRFHIPLNGCGTRYKFEDDKVVYENEIHALWTDFPPSKISRDSEFRNDMLLNINVESLTPPVASVKLGPFTLILQSYPDNSYQQPYGENEYPLVRFLRQPIYMEVRVLNRDDPNIKLVLDDCWATSTMDPDSFPQWNVVVDGCAYDLDNYQTTFHPVGSSVTHPDHYQRFDMKAFAFVSEAHVLSSLVYFHCSALICNRLSPDSPLCSVTCPVSSRHRRATGATEAEKMTVSLPGPILLLSDDSSFRGVGSSDLKASGSSGEKSRSETGEEVGSRGAMDTKGHKTAGDVGSKAVAAVAAFAGVVATLGFIYYLYEKRTVSNH | |
RUO | |
ZP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7783 | |
ZP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ZPA | |
ZP2 | |
Recombinant | |
wheat germ expression system |