missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF383 Full-length ORF (NP_689817.1, 1 a.a. - 475 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00163087-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138963

  • 508.00 € / 25µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MAEGSVMFSDVSIDFSQEEWDCLDPVQRDLYRDVMLENYGNLVSMGLYTPKPQVISLLEQGKEPWMVGRELTRGLCSDLESMCETKLLSLKKEVYEIELCQREIMGLTKHGLEYSSFGDVLEYRSHLAKQLGYPNGHFSQEIFTPEYMPTFIQQTFLTLHQIINNEDRPYECKKCGKAFSQNSQFIQHQRIHIGEKSYECKECGKFFSCGSHVTRHLKIHTGEKPFECKECGKAFSCSSYLSQHQRIHTGKKPYECKECGKAFSYCSNLIDHQRIHTGEKPYECKVCGKAFTKSSQLFQHARIHTGEKPYECKECGKAFTQSSKLVQHQRIHTGEKPYECKECGKAFSSGSALTNHQRIHTGEKPYDCKECGKAFTQSSQLRQHQRIHAGEKPFECLECGKAFTQNSQLFQHQRIHTDEKPYECNECGKAFNKCSNLTRHLRIHTGEKPYNCKECGKAFSSGSDLIRHQGIHTNK
Specifications

Specifications

NP_689817.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
81kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ35863/HSD17
ZNF383
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
163087
ZNF383 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEGSVMFSDVSIDFSQEEWDCLDPVQRDLYRDVMLENYGNLVSMGLYTPKPQVISLLEQGKEPWMVGRELTRGLCSDLESMCETKLLSLKKEVYEIELCQREIMGLTKHGLEYSSFGDVLEYRSHLAKQLGYPNGHFSQEIFTPEYMPTFIQQTFLTLHQIINNEDRPYECKKCGKAFSQNSQFIQHQRIHIGEKSYECKECGKFFSCGSHVTRHLKIHTGEKPFECKECGKAFSCSSYLSQHQRIHTGKKPYECKECGKAFSYCSNLIDHQRIHTGEKPYECKVCGKAFTKSSQLFQHARIHTGEKPYECKECGKAFTQSSKLVQHQRIHTGEKPYECKECGKAFSSGSALTNHQRIHTGEKPYDCKECGKAFTQSSQLRQHQRIHAGEKPFECLECGKAFTQNSQLFQHQRIHTDEKPYECNECGKAFNKCSNLTRHLRIHTGEKPYNCKECGKAFSSGSDLIRHQGIHTNK
RUO
ZNF383
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions