Learn More
Human ZC3HAV1 (Zinc finger antiviral protein) knockout A549 cell lysate, 1 Kit
Product Code. 32144761 Shop All Abcam Products
Product Code. 32144761 Supplier Abcam Supplier No. ab2578081Kit
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human ZC3HAV1 (Zinc finger antiviral protein) knockout A549 cell/tissue lysate.
  • Species: Human
  • Tissue: ZC3HAV1 (Zinc finger antiviral protein) knockout A549