Learn More
R&D Systems™ Human Wnt-2b (NP_078613.1) VersaClone cDNA
Specifications
Specifications
| Gene | Wnt-2 b |
| Accession Number | NP_078613.1 |
| Gene ID (Entrez) | 7482 |
| Gene Alias | Protein Wnt-13, protein Wnt-2 b, wingless-type MMTV integration site family, member 2 B, WNT13XWNT2, Xenopus, homolog of, XWNT2wingless-type MMTV integration site family, member 13 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 10 μg |