missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human WDR72 (aa 276-395) Control Fragment Recombinant Protein

Product Code. 30181901
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30181901 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30181901 Supplier Invitrogen™ Supplier No. RP98158

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-65504 (PA5-65504, PA5-63438 (PA5-63438. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Plays a major role in formation of tooth enamel (PubMed:19853237, PubMed:25008349). Specifically required during the maturation phase of amelogenesis for normal formation of the enamel matrix and clearance of enamel proteins. May be involved in localization of the calcium transporter SLC24A4 to the ameloblast cell membrane.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q3MJ13
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 256764
Name Human WDR72 (aa 276-395) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI2A3; WD repeat domain 72; WD repeat-containing protein 72; WDR72
Common Name WDR72
Gene Symbol WDR72
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GHSYIYQLLNSGLSKSIYPADGRVLKETIYPHLLCSTSVQENKEQSRPFVMGYMNERKEPFYKVLFSGEVSGRITLWHIPDVPVSKFDGSPREIPVTATWTLQDNFDKHDTMSQSIIDYF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.