Learn More
Invitrogen™ Human V-ATPase E2 (aa 155-192) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (63%), Rat (63%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62681 (PA5-62681. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q96A05 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 90423 |
| Name | Human V-ATPase E2 (aa 155-192) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4930500C14Rik; Atp6e1; ATP6EL2; Atp6v1e2; ATP6V1EL2; ATPase H+ transporting V1 subunit E2; ATPase, H transporting, lysosomal V1 subunit E2; ATPase, H+ transporting, lysosomal 31 kDa, V1 subunit E2; ATPase, H+ transporting, lysosomal 31 kDa, V1 subunit E-like 2; ATPase, H+ transporting, lysosomal V1 subunit E2; ATPase, H+ transporting, V1 subunit E-like 2; E1; lysosomal 31 kDa; testis secretory sperm-binding protein Li 235 P; vacuolar proton pump subunit E 2; vacuolar-type proton-translocating ATPase subunit E1; V-ATPase subunit E 2; VMA4; V-type proton ATPase subunit E 2 |
| Common Name | V-ATPase E2 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.