missing translation for 'onlineSavingsMsg'
Learn More

Human Tsukushi/TSK Antibody, R&D Systems™

Product Code. 18707187
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18707187 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18707187 Supplier R&D Systems Supplier No. AF3940SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Tsukushi/TSK Polyclonal specifically detects Tsukushi/TSK in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tsukushi/TSK
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8WUA8
Gene Alias E2IG4, E2IG4E2-induced gene 4 protein, leucine rich repeat containing 54, Leucine-rich repeat-containing protein 54, LRRC54, TSKtsukushi homolog, TSKU, Tsukushi, tsukushi small leucine rich proteoglycan homolog (Xenopus laevis), tsukushi, small leucine rich proteoglycan, tsukushin
Gene Symbols TSKU
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Tsukushi/TSK Thr17-Leu353 Accession # Q8WUA8
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25987
Test Specificity Detects human Tsukushi/TSK in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse TSK is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.