Learn More
Human TSSK3 (TSSK-3) knockout HeLa cell lysate, 1 Kit
Product Code. 32143709 Shop All Abcam Products
Product Code. 32143709 Supplier Abcam Supplier No. ab2634001Kit
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human TSSK3 (TSSK-3) knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: TSSK3 (TSSK-3) knockout HeLa