missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TSKS Partial ORF (NP_068379, 471 a.a. - 560 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_068379 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 60385 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16129386
|
Abnova™
H00060385-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16139386
|
Abnova™
H00060385-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This gene may play a role in testicular physiology, spermatogenesis or spermiogenesis. Expression of the encoded protein is highest in the testis and down-regulated in testicular cancer. The gene is localized to the region 19q13.3 among the related RAS viral oncogene homolog (RRAS) and interferon regulatory factor 3 (IRF3) genes, which are both involved in tumorigenesis pathways and progression. [provided by RefSeq]
Sequence: ALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDHSpezifikation
NP_068379 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TSKS1 | |
TSKS | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
60385 | |
TSKS (Human) Recombinant Protein (Q01) | |
ALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDH | |
RUO | |
TSKS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |