missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TSG-6 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2104-SP
This item is not returnable.
View return policy
Description
TSG-6 Polyclonal specifically detects TSG-6 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| TSG-6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Hyaluronate-binding protein, TNF alpha-induced protein 6, TNFAIP6, TNF-stimulated gene 6 protein, TSG6, TSG-6tumor necrosis factor alpha-inducible protein 6, TSG6tumor necrosis factor-inducible gene 6 protein, Tumor necrosis factor alpha-induced protein 6, tumor necrosis factor, alpha-induced protein 6, tumor necrosis factor-stimulated gene-6 protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7130 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P98066 | |
| TNFAIP6 | |
| Mouse myeloma cell line NS0-derived recombinant human TSG-6 Trp18-Leu277 Accession # P98066 | |
| 25 μg | |
| Primary | |
| Detects human TSG-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 70% cross-reactivity with recombinant mouse TSG-6 is observed and less than 2% cross-reactivity with recombinant human TSG-14 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction