missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TSFM Full-length ORF (NP_005717.2, 1 a.a. - 346 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010102-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16184492

  • 508.00 € / 25µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 [PubMed 17033963]).[supplied by OMIM]

Sequence: MSLLRSLRVFLVARTGSYPAGSLLRQSPQPRHTFYAGPRLSASASSKELLMKLRRKTGYSFVNCKKALETCGGDLKQAEIWLHKEAQKEGWSKAAKLQGRKTKEGLIGLLQEGNTTVLVEVNCETDFVSRNLKFQLLVQQVALGTMMHCQTLKDQPSAYSKVQWLTPVNLALWEAEAGGSLEGFLNSSELSGLPAGPDREGSLKDQLALAIGKLGENMILKRAAWVKVPSGFYVGSYVHGAMQSPSLHKLVLGKYGALVICETSEQKTNLEDVGRRLGQHVVGMAPLSVGSLDDEPGGEAETKMLSQPYLLDPSITLGQYVQPQGVSVVDFVRFECGEGEEAAETE
Specifications

Specifications

NP_005717.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COXPD3/EF-TS/EF-Tsmt
TSFM
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10102
TSFM (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSLLRSLRVFLVARTGSYPAGSLLRQSPQPRHTFYAGPRLSASASSKELLMKLRRKTGYSFVNCKKALETCGGDLKQAEIWLHKEAQKEGWSKAAKLQGRKTKEGLIGLLQEGNTTVLVEVNCETDFVSRNLKFQLLVQQVALGTMMHCQTLKDQPSAYSKVQWLTPVNLALWEAEAGGSLEGFLNSSELSGLPAGPDREGSLKDQLALAIGKLGENMILKRAAWVKVPSGFYVGSYVHGAMQSPSLHKLVLGKYGALVICETSEQKTNLEDVGRRLGQHVVGMAPLSVGSLDDEPGGEAETKMLSQPYLLDPSITLGQYVQPQGVSVVDFVRFECGEGEEAAETE
RUO
TSFM
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions